VWR® 12" Test Sieves, Half Height, Brass and Stainless Steel
Supplier: VWR International
47729-908 47729-912 47729-914 47729-918 47729-922 47729-924 47729-928 47729-932 47729-938 47729-946 47729-952 47729-954 47729-960 47729-964 47729-968 47729-972 47729-976 47729-978 47729-980 47729-982 47729-984 47729-986 47729-988 47729-990 47729-992 47729-994 47729-996 47729-998 47730-004 10811-458 10811-460 10811-520 10811-522 10811-524 10811-526 10811-610 10811-612 10811-614 10811-616 10811-780 10811-782 10811-788 10811-798 10811-806
47729-908EA 517.22 CAD
47729-908 47729-912 47729-914 47729-918 47729-922 47729-924 47729-928 47729-932 47729-938 47729-946 47729-952 47729-954 47729-960 47729-964 47729-968 47729-972 47729-976 47729-978 47729-980 47729-982 47729-984 47729-986 47729-988 47729-990 47729-992 47729-994 47729-996 47729-998 47730-004 10811-458 10811-460 10811-520 10811-522 10811-524 10811-526 10811-610 10811-612 10811-614 10811-616 10811-780 10811-782 10811-788 10811-798 10811-806
VWR® 12" Test Sieves, Half Height, Brass and Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Half Height (15/8") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Brass and stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.
Accessories information: Pans are available in half height (1-5/8") or full height (31/4") formats, in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.
Ordering information: Sieves are available in half height (15/8"), intermediate height (2") or full height (31/4"). Sieve height/depth is measured from the frame rim to the sieve cloth.