Ward's® Nutrient Broth Dehydrated Media, Powdered, IPM Scientific
Supplier: IPM SCIENTIFIC, INC.
381004
BROTH100G
470024-754EA
238.25
CAD
470024-754
470024-752
Ward's® Nutrient Broth Dehydrated Media, Powdered, IPM Scientific
Media for Microbiology
Dehydrated Media for Microbiology
Dilute quantities to any volume as needed.
- Economical alternative
- Stores much longer than prepared media
- Prepare only as much as you need at one time
Use Ward's dehydrated media for many microbiology applications. Preparation instructions are included.