Glutaraldehyde 25% in aqueous solution, EM Grade

Supplier: Electron Microscopy Sciences
Danger

Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial

16210 16221 16220 16200 16222
100503-980EA 94.64 CAD
100503-980 100503-986 100504-788 100503-978 100503-988
Glutaraldehyde 25% in aqueous solution, EM Grade
Glutaraldehyde

After glutaraldehyde has been purified to EM grade, it has a high risk of polymerizing if it is not handled properly. Electron Microscopy Sciences takes special precautions to assure a polymer-free, pure glutaraldehyde. Every lot of our EM grade aqueous glutaraldehyde is assayed prior to shipping assuring you consistent purity and the highest possible quality. If stored properly, our Glutaraldehyde has a very long shelf life.


  • Specific gravity 20/20 °C: 1.131
  • Distillation purified


EM grade aqueous glutaraldehyde, stored in volumes greater than 100 ml, with repeated defrosting and recapping, will polymerize in time, producing an absorption peak of 235 nm rather than the 280 nm of pure EM grade. The presence of polymerized glutaraldehyde greatly reduces its efficiency as a cross-linking agent, preventing proper fixation. For this reason, we provide 10 ml ampoules and 100 ml bottle quantities for larger applications.


Packaging: We package our aqueous glutaraldehyde in 10 ml, single dose ampoules, sealed under dry nitrogen, and kept refrigerated until it is shipped. We offer our glutaraldehyde in a wide range of percentages to simplify mixing procedures and avoiding waste. 1 box contains 10x10 ml ampoules.

Formula: C₅H₈O₂
MDL Number: MFCD00007025
CAS Number: 111-30-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR