Econo™ Sterile Fine Point Splinter Forceps, Sklar
Supplier: Sklar
96-2410 96-2412 96-2765
89023-754CS 590.73 CAD
89023-754 89023-756 75995-424
Econo™ Sterile Fine Point Splinter Forceps, Sklar
Forceps
Econo™ sterile splinter forceps are multipurpose thumb forceps with sharp tips used for removing small foreign objects.
  • Fine point tips
  • Ready-to-use
  • Multipurpose
Econo™ sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities. This product is straight with serrated, fine point tips.