Pierce™ Reacti-Bind™ 96-Well Polystyrene Plates, Thermo Scientific
Supplier: Thermo Scientific
15041 15042
CAPI15041PK 1087.37 CAD
CAPI15041 CAPI15042
Pierce™ Reacti-Bind™ 96-Well Polystyrene Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
96-well polystyrene flat bottom plates, treated to produce high-capacity antibody-binding characteristics for immunoassay applications.
  • Plates are certified for performance, having consistently low coefficients of variation among wells
  • Hydrophobic-ionic binding interaction
  • Corner notch for use in standard plate readers
Clear plates for colorimetric ELISA and other assays. White opaque plates for fluorescent or chemiluminescent assays.