Jumbo Syringes, Restek
Supplier: Restek
21275 21276 21277
10862-906EA 1427.48 CAD
10862-906 10862-954 10862-956
Jumbo Syringes, Restek
Syringes Syringes for Chromatography
Clear acrylic syringes, ideal for holding and dispensing large volumes of gas.
  • Ideal for large volumes of gas
  • Adjustable plunger
  • Central port with Luer lock
An adjustable plunger on the O-ring ensures that the syringe is gas-tight over a long period of time. The central port is supplied with a luer lock fitting; the secondary port is supplied with a septum nut. This enables access to the gas sample for adding standards or removing a subsample. The plunger stem is detachable, making sample storage easy.