ALTEF Gas Sampling Bags, Restek
Supplier: Restek
22958 26330 26329 26331 26328 22962 22960 22961 22963
10862-684PK 433.43 CAD
10862-684 10862-702 10862-706 10862-708 10862-710 10862-728 10862-730 10863-044 10863-046
ALTEF Gas Sampling Bags, Restek
Bags Gas Bags
ALTEF Gas Sampling Bags are an excellent low-cost alternative to Tedlar bags.
  • For collection of most VOCs
  • Very low VOC and sulfur background
  • Not recommended for ketones, acetates, hydrogen sulfide, or permanent gases
  • Durable PVDF film
  • No additives, fillers, or pigments
Sampling bags are a low-cost whole air sampling device for high level VOCs and permanent gases.
All bags feature a polypropylene combo valve with hose connection to fit ³/₁₆" Int.Ø tubing and syringe port with replaceable septum. A single eyelet provides handling convenience.