Methanizer allows ppb-level GC-FID determination of CO and CO2 without expensive equipment.
- Simple, reliable operation
- Precise temperature control
- Easy tube replacement
Precise temperature control ensures complete conversion of CO and CO2 to CH4
Low-level detection of carbon monoxide (CO) and carbon dioxide (CO2) is critical for many applications. Restek’s methanizer (CH4izer) allows ppb-level determination of CO and CO2 using a cost-effective GC-FID instead of more expensive instrumentation. The methanizer is factory set to 380 °C to ensure efficient and complete conversion of CO and CO2 to CH4, but the operator can easily adjust the temperature as desired with the touch of a button. This unit controls temperatures precisely and reliably to within ± 1 °C of the defined set point and the actual temperature is displayed in real time on an easy-to-read monitor. The Restek methanizer is less cumbersome than other models and is designed for easy installation and fast catalyst tube replacement so you spend more time analyzing samples and less time on maintenance.
Factory-set temperature (380°C) can be adjusted with just the touch of a button. Fast, easy catalyst tube replacement keeps maintenance time to a minimum.
Accessories information: Convenient installation kit (sold separately) includes all parts needed for quick installation into any Agilent GC.
Installation kit includes 1/8" Stainless steel metering gas valve, 10 foot x 1/8" 304 stainless steel gas supply line (rinsed and cleaned), 1/8" Swagelok tee union, 1/8" - 1/16" tube end reducer, 1/8" stainless steel Swagelok ball shut-off valve, MINICYL regulator with 1/8" fittings.
Delivery information: Each kit includes: Methanizer Control Box, Power Cord, Heater Chamber, Catalyst Tube, Hydrogen Supply Line, Transition Tubing, Adaptor for Agilent Capillary FID, Adaptor for Agilent Packed FID, 1/8" Swagelok Nut and Ferrules, Right-Angle Wrench.