Force FluoroPhenyl LC HPLC Columns (USP L43), Restek
Supplier: Restek
9639232 9639252 9639212 963925E 963921E 9639332 9639352 9639312 9639362 963935E 963931E 963936E 9639315 9639365 9639552 9639512 9639562 963955E 963951E 963956E 9639515 9639565 9639575
75854-234EA 1381.47 CAD
75854-234 75854-236 75854-238 75854-240 75854-242 75854-244 75854-246 75854-248 75854-250 75854-252 75854-254 75854-256 75854-258 75854-260 75854-262 75854-264 75854-266 75854-268 75854-270 75854-272 75854-274 75854-276 75854-278
Force FluoroPhenyl LC HPLC Columns (USP L43), Restek
Chromatography Columns
Restek's FluoroPhenyl phase offers chromatographers the ability to run in reversed-phase or HILIC mode for a variety of compounds.
  • Capable of both reversed-phase and HILIC separations
  • Ideal for increasing sensitivity and selectivity in LC-MS analyses
  • Offers increased retention for charged bases
The culmination of 20+ years in LC, Force performance LC columns are long-lasting and reproducible, even under the stress of UHPLC
Restek's FluoroPhenyl phase offers chromatographers the ability to run in reversed-phase or HILIC mode for a variety of compounds. Restek's FluoroPhenyl columns are also amenable to LC-MS because they are extremely reliable and efficient with acidic mobile phases.
Premium quality is backed by the strength of our 100% Pure Satisfaction guarantee. And perhaps most important, Force columns are fully scalable between 3 or 5 µm HPLC and 1.8 µm UHPLC, so you can easily transfer and optimize methods without extensive calculations.
Ordering information: Switch to a FluoroPhenyl LC column when limited retention and selectivity are observed on a C18 for basic compounds or when you need increased retention of hydrophilic compounds.