Force C18 LC HPLC Columns (USP L1), Restek
Supplier: Restek
9634362 963435E 963451E 963456E 9634515 9634565 9634575 963431E 963436E 9634232 9634252 9634212 963425E 963421E 9634332 9634352 9634312 9634315 9634365 9634552 9634512 9634562 963455E
75854-100EA 1529.3 CAD
75854-100 75854-102 75854-166 75854-168 75854-170 75854-172 75854-174 75854-312 75854-314 75854-316 75854-318 75854-320 75854-322 75854-324 75854-326 75854-328 75854-330 75854-332 75854-334 75854-336 75854-338 75854-340 75854-342
Force C18 LC HPLC Columns (USP L1), Restek
Chromatography Columns
Force C18 LC HPLC Columns are traditional end-capped C18 columns, ideal for general-purpose use in reversed-phase chromatography.
  • Compatible with moderately acidic to neutral mobile phases
  • Excellent data quality in food, environmental, bioanalytical, and more
  • Offers high hydrophobic retention
The culmination of 20+ years in LC, Force performance LC columns are long-lasting and reproducible, even under the stress of UHPLC
The general-purpose Restek C18 is a conventional monomeric octadecylsilane column suitable for analyses of a wide range of compounds from acidic through slightly basic.
Premium quality is backed by the strength of our 100% Pure Satisfaction guarantee. And perhaps most important, Force columns are fully scalable between 3 or 5 µm HPLC and 1.8 µm UHPLC, so you can easily transfer and optimize methods without extensive calculations.
Ordering information: Switch to a C18 when you need a general-purpose column for reversed-phase chromatography or when you need to increase retention of hydrophobic compounds.