Force Biphenyl LC HPLC Columns (USP L11), Restek
Supplier: Restek
9629232 9629252 9629212 962925E 962921E 9629332 9629352 9629312 9629362 962935E 962931E 962936E 9629315 9629365 9629552 9629512 9629562 962955E 962951E 962956E 9629515 9629565 9629575
75854-182EA 1353.97 CAD
75854-182 75854-184 75854-186 75854-188 75854-190 75854-192 75854-194 75854-196 75854-198 75854-200 75854-202 75854-204 75854-206 75854-208 75854-210 75854-212 75854-214 75854-216 75854-218 75854-220 75854-222 75854-224 75854-226
Force Biphenyl LC HPLC Columns (USP L11), Restek
Chromatography Columns
Force Biphenyl LC HPLC Columns are ideal for bioanalytical testing applications like drug and metabolite analyses.
  • Increased retention for dipolar, unsaturated, or conjugated solutes
  • Enhanced selectivity when used with methanolic mobile phase
  • Ideal for increasing sensitivity and selectivity in LC-MS analyses
Restek’s most popular LC stationary phase.
Columns feature heightened selectivity and retention for compounds that are hard to resolve or elute early on C18 and other phenyl chemistries. Columns also limit ionization suppression and allow simple, MS-friendly mobile phases.
The culmination of 20+ years in LC, Force performance LC columns are long-lasting and reproducible, even under the stress of UHPLC. Premium quality is backed by the strength of our 100% Pure Satisfaction guarantee. And perhaps most important, Force columns are fully scalable between 3 or 5 µm HPLC and 1.8 µm UHPLC, so you can easily transfer and optimize methods without extensive calculations.
The innovative Biphenyl is Restek's most popular LC stationary phase because it is particularly adept at separating compounds that are hard to resolve or that elute early on C18 and other phenyl chemistries. As a result, Biphenyl columns are extremely useful for bioanalytical testing applications like drug and metabolite analyses, especially those that require a mass spectrometer (MS). Increasing retention of early-eluting compounds can limit ionization suppression, and the heightened selectivity helps eliminate the need for complex mobile phases that are not well-suited for MS detection.
Ordering information: Switch to a Biphenyl when limited selectivity is observed on a C18 or if you need to increase retention of hydrophilic aromatics.