SP Wilmad-LabGlass Buchner Filter Funnels with Fritted Disc and Vacuum Adapter, SP Industries
Supplier: SP Industries
LG-7120-125 LG-7120-129 LG-7120-123
14209-294EA 681.53 CAD
14209-294 14205-766 14209-688
SP Wilmad-LabGlass Buchner Filter Funnels with Fritted Disc and Vacuum Adapter, SP Industries
Funnels Filtration Funnels Büchner Funnels
These Buchner filter funnels with vacuum adapter feature a full length Standard Taper inner joint sealed to the stem with side hose connection just above the grinding.
  • Made of borosilicate glass
With sealed-in fritted glass disc, they are available in coarse (C) and medium (M) porosities.
Recommended for use with LG-7820 heavy wall filter flasks.