CIP 220® Acid-Based Process and Research Cleaners
Supplier: Steris
CIP 220®
1D2201 1D22D3 1D2202 1D2208 1D2205
77531-094EA 25264.52 CAD
14004-192 14004-198 16200-118 77531-094 14004-194
CIP 220® Acid-Based Process and Research Cleaners
Cleaning Agents Detergents and Surface Cleansers
CIP 220® Acid detergent is a proprietary blend of hydroxyacetic (glycolic) acid, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This phosphate free low foaming detergent is ideal for use in CIP, COP and manual applications.
  • Flexibility - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Cost Savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Globally Availability - phosphate free, and manufactured with biodegradable surfactants to support harmonization of cleaning processes
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
CIP 220® Acid-based process and research cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable cleaner.