Process2Wipe® VAI® IPA70 Saturated Wipes
Supplier: Veltek Associates
VEL12-12X12-S-3030 VEL12-9X9-S-3032
76326-556CS 758.24 CAD
76326-556 77581-320
Process2Wipe® VAI® IPA70 Saturated Wipes
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Process2Wipes are saturated with 70% USP isopropyl alcohol and 30% USP water for injection.
  • 100% continuous polyester fiber is clean and absorbent
  • ISO 4 (Class 10) cleanliness
  • Validated for sterility and shelf life
These wipes are used for aseptic and non-aseptic wipe downs of filling and packaging machinery, stainless steel surfaces, Lexan, polycarbonate, glass, and any critical surfaces that require cleaning.
Packaging: Delivered with lot specific documentation. Resealable storage packaging.