Micro Surgery Scissors with Bayonet Handle, Sklar
Supplier: Sklar
98-7920 98-7924 98-7930 98-7918
10185-192EA 2298.62 CAD
10185-192 10185-194 10185-196 75996-334
Micro Surgery Scissors with Bayonet Handle, Sklar
Scissors
These micro scissors are most commonly used in iridectomy, cataract, and capsulotomy procedures.
  • Reusable
  • Nonsterile
These double spring scissors are available with various curvatures and handle lengths. The micro blade can be seen clearly under a microscope when used as a component of surgery.