Tube Furnaces with Stand for Horizontal or Vertical Operation, Model RT, Nabertherm
Supplier: NABERTHERM INC.
RT332LAN-208 RT332LAN-240 RT332OAN-208 RT332OAN-240 RT512LAN-208 RT512LAN-240 RT512OAN-208 RT512OAN-240 RT522LAN-208 RT522LAN-240 RT522OAN-208 RT522OAN-240
76538-320EA 32143.63 CAD
76538-320 76538-322 76538-324 76538-326 76538-328 76538-330 76538-332 76538-334 76538-336 76538-338 76538-340 76538-342
Tube Furnaces with Stand for Horizontal or Vertical Operation, Model RT, Nabertherm
Ovens Tube Furnaces
These compact tube furnaces are used when laboratory experiments must be performed horizontally, vertically, or at specific angles.
  • Tmax 1100, 1300 or 1500 °C
  • Compact design
  • Vertical or horizontal operation freely adjustable
  • Working height freely adjustable
  • Working tube made of C 530 ceramic
  • Type S thermocouple
  • Operation also possible separate from stand if safety guidelines are observed
  • Control system integrated in furnace base
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
The ability to configure the angle ofMore Product Information