Mini Spore Ampoules, Crosstex®
Supplier: Crosstex
OS1-50-06
10820-628EA 587.76 CAD
10820-628
Mini Spore Ampoules, Crosstex®
Ampoules
Crosstex Mini Spore ampules are self-contained biological indicators for monitoring steam processes consisting of a hermetically sealed glass ampule which contains modified Trypticase® Soy Broth (TSB) with a pH indicator and Geobacillus stearothermophilus with a population level of 106.
  • Reduced incubation time of 48 hours
  • Visually distinct positive results indicated by the media transitioning from Purple to Yellow
  • Saves time by eliminating aseptic culturing required with traditional spore strips
  • Negative controls are included in each box
The Mini Spore ampules may be utilized to monitor Steam sterilization efficacy