Iodine 0.05 M I2 (0.1N) volumetric solution, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
318981-100ML
318981-500ML
BJ318981-100MLEA
99.92
CAD
BJ318981-100ML
BJ318981-500ML
Iodine 0.05 M I2 (0.1N) volumetric solution, Fluka™
Iodine
Formula:
I₂ MW: 253.81 g/mol Boiling Pt: 184.3 Melting Pt: 114 Storage Temperature: Ambient |
MDL Number:
MFCD00011355 CAS Number: 7553-56-2 Merck Index: 13,05036 |
Specification Test Results
Assay | 0.0950 - 0.1050 N |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...