Iron(II) sulfate heptahydrate 99.5-102.0% (by manganometry), crystals, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®

Supplier: MilliporeSigma
Warning

Synonyms: Ferrous sulphate heptahydrateIron vitriolFerrous sulfate heptahydrate

1.03963.5000 1.03963.9025 1.03963.9025 1.03963.1000
CA1.03963.9025EA 4615.45 CAD
EM1.03963.5000 EM1.03963.9025 CA1.03963.9025 CA1.03963.1000
Iron(II) sulfate heptahydrate 99.5-102.0% (by manganometry), crystals, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, SAFC®
Iron (II) sulphate heptahydrate

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Certifications: Corresponds to Ph. Eur., BP, USP, FCC

Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: >60 °C (-H₂O)
Density: 1.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5

Order Now

Specification Test Results

Assay (manganometric) 99.5 - 102.0 %
Identity passes test
Appearance of solution passes test
pH-value (5 %; water) 3.0 - 4.0
Chloride (Cl) ≤ 0.01 %
Al (Aluminium) ≤ 0.01 %
As (Arsenic) ≤ 0.0002 %
Co (Cobalt) (*) ≤ 0.002 %
Cr (Chromium) ≤ 0.0050 %
Cu (Copper) ≤ 0.0050 %
Fe III (Iron III) ≤ 0.3 %
Hg (Mercury) ≤ 0.0001 %
Mn (Manganese) ≤ 0.1 %
Ni (Nickel) (*) ≤ 0.0050 %
Pb (Lead) ≤ 0.0002 %
Zn (Zinc) ≤ 0.0025 %
Residual solvents (ICH Q3C) excluded by manufacturing process

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR