Potassium chloride 99.0-100.5% (by titrimetric analysis) USP

Synonyms: Potassiumchloride

01247-10KG 01247-1KG 01247-2.5KG
CH01247-10KGEA 319.48 CAD
CH01247-10KG CH01247-1KG CH01247-2.5KG
Potassium chloride 99.0-100.5% (by titrimetric analysis) USP
Potassium chloride

Used in nutrient, gelling agent, salt substitute and yeast food.

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.984
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
Merck Index: 14,07621

Order Now

Specification Test Results

Appearance White granules
Heavy Metals ≤0.001% (as Pb)
Chloride (Cl⁻) ≤0.1%
Bromide (Br-) ≤0.1%
Iodide (I-) ≤0.005%
Sodium (Na) To pass test
Aluminium (Al) To pass test
Calcium and Magnesium To pass test
Acidity and Alkalinity To pass test
Loss on Drying ≤1% (105 °C)
Assay by titration 99.0-100.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR