Muffle Furnaces with Embedded Heating Elements in the Ceramic Muffle, Nabertherm
Supplier: NABERTHERM INC.
LM094K1LN1-208 LM094K1LN1-240 LM094K1ON1-208 LM094K1ON1-240 LM094H1LN1-208 LM094H1LN1-240 LM094H1ON1-208 LM094H1ON1-240
76538-150EA 13794.43 CAD
76538-150 76538-152 76538-154 76538-156 76538-158 76538-160 76538-162 76538-164
Muffle Furnaces with Embedded Heating Elements in the Ceramic Muffle, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
The L 9/11/SKM muffle furnaces are particularly recommended for applications that involve aggressive substances.
  • Tmax 1100 °C
  • Muffle heated from four sides
  • Furnace chamber with embedded ceramic muffle, high resistance to aggressive gases and vapours
  • Housing made of sheets of textured stainless steel
  • Optional fold down door (L models) which can be used as work platform or lift up door (LT models) with hot surface facing away from the operator
  • Adjustable working air inlet in the door
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low noise operation
The furnace has a ceramic muffle with embedded heating from four sides. The muffle furnace combines very good temperature uniformity with excellent protection of the heating elements from aggressive atmospheres. Another aspect is the smooth, nearly particle free muffle (furnace door made of fiber insulation), an important quality feature for some ashing processes.
Accessories information: Optional equipment includes: Chimney, chimney with fan or catalytic converter; Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; Protective gas connection to purge with non-flammable protective or reaction gases; Manual or automation gas supply system. Contact your Avantor representative for more information
Ordering information: These muffle furnaces with embedded heating elements can be ordered with a variety of voltage options. Models are rated at 1100 °C. Control options include the B510 controller (5 programs with 4 segments each), or the C550 controller (10 programs with 20 segments each).