Merit™ Sterile Iris Scissors, Disposable, Sklar
Supplier: Sklar
96-2506M 96-2504M 96-2505M 96-2507M
10022-486CS 1479.25 CAD
10022-486 10022-580 75995-378 75995-380
Merit™ Sterile Iris Scissors, Disposable, Sklar
Scissors
Merit™ Sterile Iris Scissors were originally designed for ophthalmic procedures, but have become a multipurpose instrument used in various surgical environments.
  • Ready-to-use
  • Multipurpose
  • Ideal for detailed work
These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used multiple procedures. Merit™ Sterile instruments are ready-to-use, sterile, disposable, office-grade instruments sold in bulk quantities.