Chest Freezer Racks for PHCbi Upright Freezers, PHC Corporation
Supplier: PHC Corporation
SCR-053-N SCR-073-N SCR-093-N SCR-053-N SCR-073-N SCR-093-N
10790-506EA 242.85 CAD
10790-506 10790-508 10790-510 CA10790-506 CA10790-508 CA10790-510
Chest Freezer Racks for PHCbi Upright Freezers, PHC Corporation
Racks Cryogenic Box Racks
These fixed side access racks hold 2" and 3" fiberboard or cardboard boxes and are for use with PHCbi upright freezers.
  • Durable and reliable
  • For long-term use
  • Locking rod included
  • Variety of sizes available
No boxes and dividers are included.
Order Now
SPECIFICATIONS
Environmentally Preferable