High-Temperature Furnaces with SiC Rod Heating, Up to 1600 °C, Nabertherm
Supplier: NABERTHERM INC.
LC034K6ON5-480 LC084K6ON8-480 LC014H6ON1-208 LC014H6ON1-240 LC034H6ON5-480 LC084H6ON8-480
76538-236EA 19819.05 CAD
76538-236 76538-238 76538-240 76538-242 76538-244 76538-246 76538-248 76538-250 76538-252 76538-254 76538-256 76538-620 76538-622 76538-624 76538-626
High-Temperature Furnaces with SiC Rod Heating, Up to 1600 °C, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These powerful laboratory muffle furnaces are available for temperatures up to 1400, 1500, 1550 or 1600 °C.
  • Tmax 1400, 1500, 1550 or 1600 °C
  • Working temperature 1500 °C (for high-temperature furnaces LHTC ../16), increased wear and tear must be expected in case of working at higher temperatures
  • Furnace chamber lined with first-class, long-life fiber material, unclassified
  • Housing made of sheets of textured stainless steel
  • Dual shell housing for low external temperatures and high stability
  • Optional fold down door (LHTC) which can be used as work platform or
More Product Information