Integrated Platform THGA Graphite Tubes, PerkinElmer
Supplier: PerkinElmer
B0504033 B3000655 B3000641 B3000653
CA97013-424PK 10872.55 CAD
CA97013-424 CA97013-428 CA97013-430 CA97013-432
Integrated Platform THGA Graphite Tubes, PerkinElmer
Tubes Graphite Tubes
These transversely-heated graphite tubes are made from a blend of high-quality base graphite material, providing the user with the required performance to yield reproducible results from day to day with respect to sensitivity (characteristic mass), accuracy and precision.
  • Integrated platform provides an even temperature heating surface
  • Significantly reduces or eliminates condensation of the sample matrix components and 'memory' effects
  • Improves the atomization efficiency for refractory elements
The graphite tube, the heart of the graphite furnace,More Product Information