Chamber Furnaces for Annealing, Hardening and Brazing, Nabertherm
Supplier: NABERTHERM INC.
N02R1NMNE-480 N02H1NMNA-208 N02H1NMNA-240 N02R1NKNE-480 N02H1NKNA-208 N02H1NKNA-240 N03R1NKNE-480 N03R1NMNE-480 N04H1NKNE-480 N04H1NMNE-480 N05H1NMNE-480 N05H1NKNE-480 N06H1NKNE-480 N06H1NMNE-480 N01H1NMNA-208 N01H1NMNA-240 N01H1NKNA-208 N07H1NMNE-480 N07H1NKNE-480
76538-198EA 13531.82 CAD
76538-198 76538-200 76538-202 76538-204 76538-206 76538-208 76538-210 76538-212 76538-214 76538-216 76538-218 76538-220 76538-222 76538-224 76538-226 76538-228 76538-230 76538-232 76538-234
Chamber Furnaces for Annealing, Hardening and Brazing, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
The N 7/H - N 87/H chamber furnaces are designed to withstand harsh use in the laboratory.
  • Three-sided heating from both sides and the bottom, heating elements on support tubes ensure free heat radiation and a long service life
  • Bottom heating protected by heat-resistant SiC plate, low energy consumption due to multi-layer insulation
  • Exhaust opening in the side of the furnace, or on back wall of chamber furnace for model N 31/H and higher
  • Base frame included in the delivery, N 7/H - N 17/HR designed as benchtop model
  • Parallel guided downward swinging door (user protected from heat radiation)
  • Gas spring dampers provide for easy door opening and closing
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
The furnaces can be extended with a variety of accessories, like annealing boxes for operation under protective gas, roller guides, or a cooling station with a quench tank. Even high-performance applications such as the annealing titanium in medical applications can be implemented without the use of expensive and complicated annealing systems.
The chamber furnaces are ideal when heat-treating metals and robust insulation with light refractory bricks is necessary.
Accessories information: The wide selection of chamber furnaces for annealing, hardening and brazing can be extended with a variety of accessories to suit your application. The accessories detailed here represent only a small fraction of the products available. For further details, please see our separate catalogues for heat-treatment furnaces and hardening accessories. Optional accessories include annealing boxes; annealing tray with holder; hearth plates; hardening tongs; heat treating foil; gloves. Contact your Avantor representative for more information.
Ordering information: These chamber furnaces can be ordered with a variety of sizes and voltage options. Models are rated at 1280 °C. Control options include the B500 controller (5 programs with 4 segments each), or the C540 controller (10 programs with 20 segments each).