High-Temperature Furnaces with MoSi₂ Heating Elements, Up to 1800 °C, LHT Model, Nabertherm
Supplier: NABERTHERM INC.
LHT226VN-208 LHT226VN-240 LHT426VN-480 LHT826VN-480 LHT227VN-208 LHT227VN-240 LHT228VN-208 LHT228VN-240 LHT427VN-480 LHT428VN-480 LHT827VN-480 LHT828VN-480 LHT127WN1-208 LHT127WN1-240 LHT327VN1-208 LHT327VN1-240
76538-258EA 28809.01 CAD
76538-258 76538-260 76538-262 76538-264 76538-270 76538-272 76538-274 76538-276 76538-278 76538-280 76538-282 76538-284 76538-286 76538-288 76538-290 76538-292
High-Temperature Furnaces with MoSi₂ Heating Elements, Up to 1800 °C, LHT Model, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
Designed as tabletop models, these compact high-temperature furnaces have a variety of advantages.
  • Tmax 1600, 1750, or 1800 °C
  • High-quality molybdenum disilicide heating elements
  • Furnace chamber lined with first-class, long-life fiber material, unclassified
  • Housing made of sheets of textured stainless steel
  • Dual shell housing with additional fan cooling for low surface temperature
  • Compact design with lift door, opening upwards
  • Adjustable air inlet
  • Exhaust air opening in the roof
  • Type B thermocouple
  • Switchgear with steady control of the heating elements
More Product Information