Revised GPC Calibration Mix (5 components), Restek
Supplier: RESTEK CORP
32041
32042
RK32041EA
52.14
CAD
RK32041
RK32042
Revised GPC Calibration Mix (5 components), Restek
Standards
Organic Standards
Contains: Bis(2-ethylhexyl) phthalate (117-81-7), 5 mg/mL; Corn oil (8001-30-7), 250 mg/mL; Methoxychlor (72-43-5), 1.0 mg/mL; Perylene (198-55-0), 0.2 mg/mL; Sulfur (7704-34-9), 0.8 mg/mL