DW-VOC Mix #2 (12 components), Restek
Supplier: RESTEK CORP
30038
30220
RK30038EA
68.73
CAD
RK30038
RK30220
DW-VOC Mix #2 (12 components), Restek
Standards
Environmental Analysis Standards
Contains: Chlorobenzene (108-90-7); 1,2-Dichlorobenzene (95-50-1); cis-1,2-Dichloroethene (156-59-2); trans-1,2-Dichloroethene (156-60-5); 1,2-Dichloropropane (78-87-5); Ethylbenzene (100-41-4); Styrene (100-42-5); Tetrachloroethene (127-18-4); Toluene (108-88-3); m-Xylene (108-38-3); o-Xylene (95-47-6); p-Xylene (106-42-3)