Sterility Testing Medium-A, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M017-500G M017-2.5KG M017-5KG
CA95020-050EA 545.91 CAD
CA95020-050 CA89222-218 CA89222-220
Sterility Testing Medium-A, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
This sterility testing medium A is used for checking presence of aerobic microorganisms in pharmacopoeial articles.
The medium contains peptic digest of animal tissue and meat extract which provide carbonaceous, nitrogenous compounds and essential growth nutrients for the growth of microorganisms. Sodium chloride maintains the osmotic balance.