Chamber Furnaces with Brick Insulation or Fiber Insulation, Nabertherm
Supplier: NABERTHERM INC.
LH1232NKNE-480 LH1232NMNE-480 LH1242NKNE-480 LH1242NMNE-480 LH0232NMNE-480 LH0332NMNE-480 LH0342NKNE-480 LH0342NMNE-480 LH0632NMNE-480 LH0642NKNE-480 LH0642NMNE-480 LH2122NKNE-480 LH2122NMNE-480 LH2132NKNE-480 LH2132NMNE-480 LF0232NKNE-480 LF0332NKNE-480 LF0632NKNE-480 LF1232NKNE-480 LF1232NMNE-480 LF1242NKNE-480 LF1242NMNE-480 LF0232NMNE-480 LF0242NKNE-480 LF0242NMNE-480 LF0332NMNE-480 LF0342NKNE-480 LF0342NMNE-480 LF0632NMNE-480 LF0642NKNE-480 LF0642NMNE-480 LH0222NKNE-480 LH0232NKNE-480 LH0242NKNE-480 LH1222NMNE-480 LH0222NMNE-480 LH0242NMNE-480 LH2142NKNE-480 LH2142NMNE-480 LH0322NMNE-480 LH0332NKNE-480 LH0622NMNE-480 LH0632NKNE-480 LH0322NKNE-480
76538-058EA 32632.1 CAD
76538-058 76538-060 76538-062 76538-064 76538-066 76538-068 76538-070 76538-072 76538-074 76538-076 76538-078 76538-080 76538-082 76538-084 76538-086 76538-092 76538-094 76538-096 76538-098 76538-100 76538-102 76538-104 76538-106 76538-108 76538-110 76538-112 76538-114 76538-116 76538-118 76538-120 76538-122 76538-124 76538-126 76538-128 76538-130 76538-132 76538-134 76538-136 76538-138 76538-140 76538-142 76538-144 76538-146 76538-148
Chamber Furnaces with Brick Insulation or Fiber Insulation, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
The LH 15/12 - LF 120/14 chamber furnaces have been trusted for many years as professional chamber furnaces for the laboratory.
  • Tmax 1200, 1300, or 1400 °C
  • Dual shell housing with rear ventilation, provides for low shell temperatures
  • Five-sided heating for very good temperature uniformity
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Controller mounted on furnace door and removable for comfortable operation
  • Protection of bottom heating and flat stacking surface provided by embedded SiC plate in the floor
  • LH models:
More Product Information