Exempted Drug of Abuse Reference Standards, Restek
Supplier: RESTEK CORP
34000 34002 34004 34005 34006 34007 34008 34009 34015 34017 34021 34024 34025 34027 34029 34032 34034 34036 34037 34038 34066 34071 34078 34079 34081 34082 34083 34084 34085 34086 34042 34044 34046 34047 34049 34050 34051 34053 34054 34055 34056 34057 34059 34062 34063 34064 34065 34010 34011 34014
75835-236EA 112.73 CAD
75835-236 75835-238 75835-240 75835-242 75835-244 75835-246 75835-248 75835-250 75835-252 75835-254 75835-256 75835-258 75835-260 75835-262 75835-264 75835-266 75835-268 75835-270 75835-272 75835-274 75835-276 75835-278 75835-280 75835-282 75835-284 75835-286 75835-288 75835-290 75835-292 75835-294 75835-296 75835-298 75835-302 75835-306 75835-308 75835-310 75835-312 75835-314 75835-316 75835-318 75835-320 75835-322 75835-324 75835-900 75835-902 75835-904 75835-906 75835-908 75835-910 75835-912 75835-914 75835-916 75835-918 75835-920 75835-922 75835-924 75835-926 75835-928 75835-930 75835-934 75835-936 75835-938 75835-940 76124-750 76124-752 76124-754
Exempted Drug of Abuse Reference Standards, Restek
Standards Organic Standards
Caution: Note: Stressed shipping studies indicate that this product remains stable after being continuously exposed to an elevated temperature of 40 °C for 7 days. Ground freight is the default shipping method for this product; expedited methods are available upon request.