Single-Component Explosives Solutions, Restek
Supplier: RESTEK CORP
31657 31658 31662 31669 31498 31600 31671 31452 31453 31499 31601 31602 31661 31663 31664 31665 31666 31667 31668
RK31657EA 56.09 CAD
RK31657 RK31658 RK31662 RK31669 RK31498 RK31600 RK31671 75834-694 75834-696 75835-696 75835-698 75835-700 75835-760 75835-762 75835-764 75835-766 75835-768 75835-770 75835-772
Single-Component Explosives Solutions, Restek
Standards Organic Standards
  • Supports US Department of Defense base closures and remediation
  • Mixtures and singles supporting LC US EPA Method 8330
  • Mixtures and singles supporting GC-ECD US EPA Method 8095
  • Internal standards and surrogates to support both methods