QuEChERS Single-Component Reference Standards, Restek
Supplier: RESTEK CORP
33257 33258 33259
75835-194EA 52.09 CAD
75835-194 75835-196 75835-198 75835-200 75835-204 75835-206
QuEChERS Single-Component Reference Standards, Restek
Standards Organic Standards
  • Ready to use for QuEChERS extractions—no dilutions necessary
  • Support for GC and LC with MS, MS/MS, and selective detectors