VWR® 12" Test Sieves, Half Height, All Stainless Steel
Supplier: VWR International
47730-488 47730-506 47730-508 47730-512 47730-514 47730-516 47730-520 47730-522 47730-524 47730-526 47730-528 47730-530 47730-532 47730-534 47730-536 47730-538 47730-540 47730-542 47730-544 47730-546 47730-548 47730-550 47730-552 47730-554 47730-556 47730-562 47730-564 47730-568 47730-570 47730-574 10811-592 10811-594 10811-596 10811-598 10811-600 10811-602 10811-604 10811-606 10811-608 10811-726 10811-728 10811-730 10811-732 10811-786
47730-488EA 712.63 CAD
47730-488 47730-506 47730-508 47730-512 47730-514 47730-516 47730-520 47730-522 47730-524 47730-526 47730-528 47730-530 47730-532 47730-534 47730-536 47730-538 47730-540 47730-542 47730-544 47730-546 47730-548 47730-550 47730-552 47730-554 47730-556 47730-562 47730-564 47730-568 47730-570 47730-574 10811-592 10811-594 10811-596 10811-598 10811-600 10811-602 10811-604 10811-606 10811-608 10811-726 10811-728 10811-730 10811-732 10811-786
VWR® 12" Test Sieves, Half Height, All Stainless Steel
Sieves Test Sieves
VWR® 12" Diameter (304.8 mm ) all stainless steel test sieves are durable and ideal for sieving applications where a larger sample size is required.
  • Half Height (31/4") sieves
  • Highly durable
  • Ideal for sieving applications requiring a larger sample size
  • Stainless steel construction
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying the sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes, and Centerline® sieve inspection or calibrationMore Product Information