AccuGreen™ High Sensitivity and Broad Range dsDNA Quantitation Kits, Biotium
Supplier: Biotium
31066 31066-T 31069 31069-T 99820-T 99838-T
75845-716EA 577.02 CAD
75845-716 75845-718 76204-710 76204-712 76414-986 76414-988
AccuGreen™ High Sensitivity and Broad Range dsDNA Quantitation Kits, Biotium
Assays Cellular Assays
The AccuGreen™ High Sensitivity and Broad Range dsDNA quantitation kits are designed for use with handheld fluorometers such as the Qubit® fluorometers from Thermo Fisher Scientific.
  • Designed specifically for Thermo’s Qubit® fluorometer
  • Linear range: 0.1 to 100 ng dsDNA (AccuGreen™ High Sensitivity), 2 to 1000 ng dsDNA (AccuGreen™ Broad Range)
  • Sample concentration range: 5 pg/µl to 100 ng/µl (AccuGreen™ High Sensitivity), 0.1 to 1000 ng/µl (AccuGreen™ Broad Range)
  • Green fluorescence emission
  • Replaces Qubit® dsDNA HS and BR assay kits for cost savings
Fluorescent dsDNA quantitation kits for sensitive and selective detection of purified dsDNA samples on handheld fluorometers such as the Qubit® fluorometer. Kits available with or without DNA standards, for users that wish to use their own standards.