BoneClones® Juvenile Human Skulls
Supplier: BONE CLONES HOLDINGS
BC-111 BC-275 BC-277 BC-135 BC-270 BC-301
470227-966EA 569.1 CAD
470227-966 470227-968 470227-970 470227-972 470227-974 470227-976 470227-978 470227-980
BoneClones® Juvenile Human Skulls
Models Human Biology Models
How does the skull change as we age?
  • Compare multiple ages to show skull development
  • High quality casts of actual specimens
  • Material resists warping and cracking
These skulls show growth and development from 15 months to adolescence. As race and sex can not be reliably determined until adulthood, these skulls are a great way to examine the changes as an individual matures. Each skull has a separate jaw, and show the dentition of the individual at the approximate age. 2 of the skulls have had the dentition exposed (5 Year, and 12 Year) showing the placement of the permeant teeth.
Ordering information: Each skull is shipped with an osteological evaluation report. Skulls with 3 parts include a removable calvarium.