SP Wilmad-LabGlass Mobile Phase Filtration/Degassing Cap, SP Industries
Supplier: SP Industries
BP-1915-047 BP-1914-001
14208-546EA 1757.39 CAD
14208-546 14202-654
SP Wilmad-LabGlass Mobile Phase Filtration/Degassing Cap, SP Industries
Closures Safety Caps and Safety Waste Caps
Filtration/Degassing caps are constructed of PTFE with polypropylene vacuum adapter for 1/4" I.D
Lower screw cap has 45 mm thread and upper screw clamp holds mobile phase pickup adapter. Caps include a removable coarse fritted glass filter support (specially treated to resist flaking) and a Viton® O-ring for sealing to the reservoir.
tubing.