Polystyrene Reagent Reservoirs
Supplier: MTC BIO, INC.
P8010_WARDS P8025_WARDS P8050_WARDS P8051_WARDS P8100_WARDS P8010-1S_WARDS P8025-1S_WARDS P8050-1S_WARDS P8051-1S_WARDS P8100-1S_WARDS P8010-5S_WARDS P8025-5S_WARDS P8050-5S_WARDS P8051-5S_WARDS P8100-5S_WARDS
470228-448CS 215.05 CAD
470228-448 470228-450 470228-452 470228-454 470228-456 470228-458 470228-460 470228-462 470228-464 470228-466 470228-468 470228-470 470228-472 470228-474 470228-476
Polystyrene Reagent Reservoirs
Reagent Reservoirs
Standard 4-channel, 8-channel, 12-channel and various special purpose pipettes are all compatible with these disposable, polystyrene reservoirs. To facilitate easy, dripless pouring, all versions have independent spouts in each corner. Bottoms are formed with a sharply tapered 'V' shape to minimize waste and reagent consumption.
  • Graduated
  • Anti-static
  • Sharp "V" Bottoms
  • Pouring spout on all 4 corners
  • Divided options are available
  • All reservoirs are RNAse- and DNAse-free