Dewar Condensers, Chemglass
Supplier: Chemglass
CG-1209-A-20 CG-1209-A-27 CG-1209-A-28 CG-1209-A-30 CG-1209-A-22 CG-1209-A-26
80067-244EA 495.68 CAD
80067-244 80067-256 80067-258 80067-260 80067-248 80067-254
Dewar Condensers, Chemglass
Condensers
Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen.
Condenser has an outer standard taper joint set at a 75° angle to the condenser body and a lower inner drip tip joint, both being the same size.