Direct red 80 kit stain for histology

Supplier: POLYSCIENCES INC-RESEARCH CHEM

Synonyms: CI 35780, Sirius Red F3B, Sirius Red, Picro-Sirius Red

24901-250 24901-500
75805-978KT 598.27 CAD
75805-978 75805-980
Direct red 80 kit stain for histology
Direct Red 80

This stain binds specifically to collagen fibrils of varying diameter to distinguish collagen type I from type III


This stain helps in quantifying the amount of collagen in a given area of myocardial tissue and is used to analyse the quantitative aspect of liver fibrosis.


Delivery information: Kit contains Phosphomolybdic Acid, Picrosirius Red F3BA Stain, 0.1 N Hydrochloride Acid.

Formula: C₄₅H₂₆N₁₀Na₆O₂₁S₆
MDL Number: MFCD00054389
CAS Number: 2610-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR