Mag-Bind® cfDNA Kit, Omega Bio-Tek
Supplier: OMEGA BIO-TEK CA
M3298-00 M3298-01 M3298-02
CA76035-972EA 135.69 CAD
CA76035-972 CA76035-974 CA76036-024
Mag-Bind® cfDNA Kit, Omega Bio-Tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Mag-Bind® cfDNA Kit is designed for rapid and reliable isolation of circulating DNA from 500 - 4 000 µL plasma/serum samples.
  • Designed for rapid isolation
  • Can be processed manually or with automated platforms
  • Eliminates additional steps
  • Hands-free operation
The Mag-Bind® cfDNA Kit can be processed manually or with automated platforms
The uniquely formulated binding buffer allows for large sample volumes to be processed in automated formats with 4 mL of serum or plasma being processed in 24-well plates. The magnetic properties of the Mag-Bind® Particles CH enable fast magnetic separation, even when using large volumes. The high binding capacity of the beads allows for lower volumes of magnetic particles needed, thus reducing the final elution volume required. 4 mL of serum or plasma can be eluted in as low as 50 µL.
The system combines the reversible nucleic acid-binding properties of Mag-Bind® paramagnetic particles with a unique binding system that targets smaller DNA fragments (150 - 400 bp) and minimizes the binding of larger fragments, such as gDNA.
The procedure eliminates the need for funnels and vacuum steps, providing hands-free operation in automated protocols.