Sodium hydroxide solution 32% (by acidimetry), EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Supplier: EMD MILLIPORE – PCS CA EXCEP
|
Danger
|
Synonyms:
Caustic soda
Filtered through a 0.2 µm filter.
This product has been manufactured from Sodium hydroxide pellets Ph Eur, BP, JP, NF and water for injection (wfi) acc. USP which conforms with highly purified water (hpw) acc. Ph Eur.
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Formula:
NaOH MW: 40 g/mol Boiling Pt: 1390 Melting Pt: 323 Density: 1.35 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1824 ADR: 8 Merck Index: 14,08627 |
Specification Test Results
Assay (acidimetric,NaOH) | 32.0 - 33.0 % |
Carbonate (as Na₂CO₃) | ≤ 1.0 % |
Chloride (Cl) | ≤ 5 ppm |
Sulfate (SO₄) | ≤ 30 ppm |
Heavy metals (as Pb) | ≤ 5.0 ppm |
Al (Aluminium) | ≤ 10 ppm |
Fe (Iron) | ≤ 3 ppm |
Ni (Nickel) * | ≤ 2.00 ppm |
Residual solvents (ICH Q3C) | ...excluded by production process |
Endotoxins | ≤ 2.0 I.U./ml |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...