Wards® Luria Broth Powdered Media
Supplier: ALDON CORP SE
AD-MM1025-1KG AD-MM1025-100G AD-MM1025-500G
470227-484EA 285.35 CAD
470227-484 470227-504 470227-524
Wards® Luria Broth Powdered Media
Media for Microbiology Dehydrated Media for Microbiology
High quality microbiological media at an economical price.
  • SDS forms available for each media type
  • Carefully blended for consistency and reliability
  • Longer shelf life than prepared media
  • Economical alternative
Use's Ward's® own dehydrated media for many of your routine microbiology applications.
Combine powder with water according to the mixture instructions below and sterilize for ready-to-use bacterial media.