Drum and Carboy Funnels, Chemglass
Supplier: Chemglass
CG-1766-02 CG-1766-03 CG-1766-01
89028-778EA 110.97 CAD
89028-778 89028-780 89028-776
Drum and Carboy Funnels, Chemglass
Funnels Filtration Funnels Drum Funnels
Large funnels with handles are ideal for filling columns, barrels, tanks, and carboys providing ample volume and fast flow
The two smaller sizes have a handle at the rim. Molded of natural polypropylene, funnels are autoclavable.