Chain Clamps, Chemglass
Supplier: Chemglass
CG-9220-01 CG-9220-02
80063-614EA 173.52 CAD
80063-614 80063-616
Chain Clamps, Chemglass
Clamps General Purpose Clamps
Small and large size chain clamps accommodate up to 170mm (61/2") O.D
  • Grip is vinyl coated
  • Quick slip-on chain connection has large, easy-to-turn adjusting knob
and 290mm (111/2") O.D. respectively.