Pops® DPD Free Chlorine Reagent Powder, HF Scientific
Supplier: HF SCIENTIFIC INC
10300
10301
10306
10307
10312
66130-400EA
376.75
CAD
66130-400
66130-402
66130-408
66130-410
66130-416
Pops® DPD Free Chlorine Reagent Powder, HF Scientific
Environmental Test Kits
For determination of free chlorine by the DPD method
This individually packaged doses of reagent used to deliver a measured amount of DPD Reagent to a 5, 10 or 25 mL testing sample.
Ordering information: Pack of 200 or 1000 units.