Brain Heart Infusion (BHI) Broth, Dry Media, Teknova
Supplier: TEKNOVA CA
B9500 B9505
CA101319-540EA 807.66 CAD
CA101319-540 CA101324-842
Brain Heart Infusion (BHI) Broth, Dry Media, Teknova
Media for Microbiology Dehydrated Media for Microbiology
Brain Heart Infusion Broth is a general-purpose medium suitable for the cultivation of a wide variety of organisms, including bacteria, yeasts and molds.
  • Cultivates a wide variety of organisms
  • Made of 3.7% Brain Heart Infusion Powder
Ambient shipping conditions. Store at room temperature.