Delong Shaker Flasks with Three Extra Deep Side Baffles, Chemglass
Supplier: Chemglass
CLS-2046-01 CLS-2046-02 CLS-2046-03 CLS-2046-05 CLS-2046-06 CLS-2046-08 CLS-2046-10 CLS-2046-04
30623-234EA 127.54 CAD
30623-234 30623-236 30623-238 30623-240 30623-242 30623-244 30623-248 76380-146
Delong Shaker Flasks with Three Extra Deep Side Baffles, Chemglass
Flasks Erlenmeyer Flasks
Extra deep baffle indents on the lower outside edge of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker.
  • Shaker flasks are made of Class A borosilicate glass Type 1
Delong style neck is made to fit stainless steel or plastic closures, while the longer neck reduces the amount of splashing that would reach the closure.