Total Sugar Test Strips, EMD Millipore
Supplier: MilliporeSigma
1.16136.0001 1.16136.0001
CA1.16136.0001EA 505 CAD
CA1.16136.0001 EM1.16136.0001
Total Sugar Test Strips, EMD Millipore
Environmental Test Kits
Total Sugar Test can be used to monitor glucose and fructose in food, such as potatoes, and in fruit juices, and also optimize your wine making process.